Specifications
- Description:PhiX174 RF II DNA
- Size:150 µg
- Cat. no.:CA101228-454
Specifications
About this item
This is the double-stranded, nicked, circular form of ΦX174 DNA (relaxed form)
The molecular weight of ΦX174 DNA is 3.50 X 106 daltons it is 5,386 base pairs in length. Greater than 90% of the molecules are RF II, the remainder are RF I (supercoiled) and linear ΦX174 DNA. DNASU and Addgene are central repositories for plasmid clones and collections that may also be helpful. ΦX174 RF II is prepared by relaxing (nicking) ΦX174 RF I DNA in high salt at 75°C. Greater than 90% of the molecules are in the RF II form as indicated by agarose gel electrophoresis.