O-Glycosidase
Supplier: New England Biolabs (NEB)
Certificates
About this item
O-Glycosidase, also known as Endo-α-N-Acetylgalactosaminidase, catalyzes the removal of Core 1 and Core 3 O-linked disaccharides from glycoproteins.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.