NEBNEXT® End Repair Reaction Buffer, New England Biolabs
Catalog #: CA101710-270
Supplier: New England Biolabs (NEB)
CAS Number:
Specifications
- Description:NEBNext End Repair Reaction Buffer
- Size:2.0 mL
- Cat. no.:CA101710-270
Specifications
About this item
New England Biolabs supplies a 10X reaction buffer for use with NEBNext® End Repair Enzyme Mix
At a 1X concentration this reaction buffer assures optimal activity of the enzyme mix.
Customers Who Bought This Also Bought