ContactUsLinkComponent
LONGAMP® Taq PCR Kit, New England Biolabs
LONGAMP® Taq PCR Kit, New England Biolabs
Catalog #: CA101444-720
CAS Number:  
LongAmp* Taq PCR Kit 100 reactions
LONGAMP® Taq PCR Kit, New England Biolabs
Catalog #: CA101444-720
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    LongAmp Taq PCR Kit
  • Size:
    100 Reactions
  • Cat. no.:
    CA101444-720
Specifications
About this item
The LongAmp Taq PCR Kit features LongAmp Taq DNA Polymerase, which is a unique blend of Taq and Deep VentR ™ DNA Polymerases
The 3´→5´ exonuclease activity of Deep VentR Polymerase increases the fidelity and robust amplification of Taq DNA Polymerase. LongAmp Taq DNA Polymerase offers two fold higher fidelity than Taq Polymerase alone. The LongAmp Taq PCR Kit is supplied with LongAmp Taq Reaction Buffer, which is optimized to give highest PCR yields up to 30 kb. The kit can generate a wide range of PCR products, up to 30 kb from plasmid or from human genomic DNA.
Customers Who Bought This Also Bought