Endo H
Supplier: New England Biolabs (NEB)
Certificates
About this item
Endoglycosidase H is a recombinant glycosidase which cleaves within the chitobiose core of high mannose and some hybrid oligosaccharides from N-linked glycoproteins.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.