ContactUsLinkComponent
E.coli Ribosome, New England Biolabs
E.coli Ribosome, New England Biolabs
Catalog # CA101641-600
undefined
E.coli Ribosome, New England Biolabs
Catalog # CA101641-600
Supplier Number:  P0763S
Specifications
  • Description:
    E.coli Ribosome
  • Size:
    1 mg
  • Cat. no.:
    CA101641-600
Specifications
About this item
The ribosome is the machinery for protein synthesis in a cell
The 70S E. coli Ribosome consists of a small subunit (30S) and a large subunit (50S). This preparation of ribosomes is highly active in NEB’s PURExpress Protein Synthesis Kit, and can be used in ribosome structure and function studies, as a target for drug screening and as starting material for isolation of native ribosomal RNAs (5S, 16S, 23S). E. coli Ribosome is supplied as a 33.3 mg/ml solution.