E. coli DNA Ligase, New England Biolabs
Supplier: New England Biolabs (NEB)
Certificates
About this item
Catalyzes the formation of a phosphodiester bond between juxtaposed 5'-phosphate and 3'-hydroxyl termini in duplex DNA containing cohesive ends.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.