Chitin Magnetic Beads, New England Biolabs
Catalog #: CA101227-556
Supplier: New England Biolabs (NEB)
Specifications
- Description:Chitin Magnetic Beads
- Size:5 mL
- Cat. no.:CA101227-556
Specifications
About this item
An affinity matrix for the small-scale isolation of target proteins fused to a chitin binding domain (CBD)
Chitin beads have been prepared having a magnetite core. This permits the magnetic isolation of CBD-fusion proteins from cell culture supernatants; after which, the matrix can be regenerated without loss of binding capacity. Immobilized fusion proteins can be used in subsequent experiments to capture target proteins from crude cell lysates that interact with the immobilized fusion protein.
Customers Who Bought This Also Bought