About this item
Bsu DNA Polymerase I, Large Fragment retains the 5´→ 3´ polymerase activity of the Bacillus subtilis DNA polymerase I, but lacks the 5´→ 3´ exonuclease domain
This large fragment naturally lacks 3´→ 5´ exonuclease activity.