Specifications
- Chemical method:Phthalein complexone
- Description:Test with Strips
- Cat. no.:CA1.16997.0001
- Range:0,1 - 30,0 °d (1 - 300 mg/l CaO)
- Supplier No.:1.16997.0001
- No. of Tests:50 strips
Specifications
About this item
Test strips for the determination of water hardness with reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.
- Test kits with reagents for reliable documented quantitative results
- Barcode calibration
- Low analysis cost
- Test strips with minute amount of chemicals for easy disposal
The test measures the content of salts of the alkaline earth metals calcium, magnesium, strontium and barium in groundwater and surface water, drinking water, mineral water and boiler water.