Specifications
- Chemical method:Ba-thorin complex
- Description:Test with Strips
- Cat. no.:CA1.10019.0001
- Range:200-400-800-1200-1600 mg/l SO₄
- Supplier No.:1.10019.0001
- No. of Tests:100 strips
Specifications
About this item
The sulfate test can be used to give a rapid estimation of the sulphate ion content of drinking and industrial water, as well as of effluent discharged, for instance, from electroplating works and leather goods manufacturers.
MilliporeSigma # 1.10019.0001 replaces and is identical to MilliporeSigma # 10019-1