Glass Plates, Cytiva
Catalog #: CA95040-384
Supplier: Cytiva
Specifications
- Cat. no.:CA95040-384
- L×W×H:125×260×1 mm
- Supplier No.:80-1106-29
Specifications
About this item
Customers Who Bought This Also Bought
Extran® MN 01 Powdered Detergent for Manual Washing, MilliporeSigma
Supplier: MilliporeSigma
214,10 $ to 1 039,32 $
Water, ARISTAR® ULTRA, Ultrapure for trace metal analysis, VWR Chemicals BDH®
Supplier: BDH
334,23 $ to 2 005,34 $
Acetate Buffer Solution, pH 4.0, VWR Chemicals BDH®
Supplier: VWR International
270,18 $ to 1 066,09 $
Tryptic Soy Agar (TSA), Soybean Casein Digest Agar, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
39,00 $
Methanol-[D₄] ≥99.8% (Isotopic), MagniSolv™ for NMR spectroscopy, Sigma-Aldrich®
Supplier: MilliporeSigma
374,70 $
Chlorine Standard Solution, 50 to 75 mg/L as Cl₂, 10 ml Voluette Ampules (NIST), Hach
Supplier: Hach
176,20 $