DEAE Sephadex™ A-25 Ion Exchange Chromatography Media
Supplier: Cytiva
Certificates
About this item
DEAE Sephadex™ A-25 and DEAE Sephadex™ A-50 are weak anion exchangers. Sephadex™ ion exchangers are produced by introducing functional groups onto the cross-linked dextran matrix. These groups are attached to glucose units in the matrix by stable ether linkages.
- Suitable for batch techniques
- High binding capacities
- Well-proven and widely used especially in approved plasma fractionation processes (DEAE Sephadex™ A-50 only)
Different types of ion exchangers (DEAE, QAE and CM) and two different porosities (A-25/C-25 and A-50/C-50) are available.
These groups are attached to glucose units in the matrix by stable ether linkages.
Related Information
Certificates
Search by Lot Number17-0170-01_42 DF.pdf 17-0170-02_42_DF.pdf 17-0170-02_42_DF.pdf 17017001_DATA FILE.pdf 17017002_DATA FILE.pdf
Datasheet
17-0170-01_42 IN.pdf 17-0170-02_42 MS.pdf 17-0170-02_42_IN.pdf 17017001_INSTRUCTIONS.pdf 17017002_INSTRUCTIONS.pdf
Manual
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
Customers Who Bought This Also Bought