DEAE Sepharose™ CL-6B Ion Exchange Chromatography Media, Cytiva
Catalog #: CA95016-964L
Supplier: Cytiva
CAS Number:
Specifications
- Particle size:90 µm
- pH range:3 - 9
- Description:DEAE Sepharose™ CL-6B
- Flow rate:<150 cm/h
- Size:500 mL
- Matrix:6% Cross-linked agarose
- Ligand:Diethylaminoethyl
- Storage temperature:4 - 30 °C
- Binding capacity:Thyroglobulin (MW 669 000) 2 mg/mL, HSA (MW 68 000) 170 mg/mL, alpha-lactalbumin (MW 14 300) 150 mg/mL
- Total ionic capacity:0,13 - 0,17 mmol/ml
- Cat. no.:CA95016-964L
- Type:Liquid
- Supplier No.:17-0710-01
Specifications
About this item
DEAE Sepharose™ CL-6B is a weak anion exchanger.
- Excellent flow properties
- High capacity for proteins of all pI values
The ion exchange group is diethylaminoethyl which remains charged and maintains consistently high capacities over the entire working range, pH 3 to 9. Stable to all commonly used aqueous buffers additives such as 8 M urea and 6 M guanidine hydrochloride.
Customers Who Bought This Also Bought