Capto™ Butyl Hydrophobic Interaction Chromatography Media, Cytiva
Catalog #: CA97067-926
Supplier: Cytiva
CAS Number:
Specifications
- Particle size:75 µm
- pH range:3 - 13
- Description:Capto™ Butyl
- Size:25 mL
- Matrix:Highly cross-linked agarose
- Ligand:Butyl
- Storage temperature:4 - 30 °C
- Binding capacity:27 mg BSA/ml
- Cat. no.:CA97067-926
- Type:Liquid
- Supplier No.:17545901
Specifications
About this item
Capto Butyl is a hydrophobic interaction chromatography (HIC) media used in the capture and intermediate stages of protein purification, Their combination of high capacity, high flow rate and low backpressure reduces process cycle times.
- Improved productivity and process economy in downstream operations
- Very high flow rates and large sample volume processing
- Excellent chemical stability
Customers Who Bought This Also Bought