Capto™ Phenyl (High Sub) Hydrophobic Interaction Chromatography Media, Cytiva
Catalog #: CA97067-924
Supplier: Cytiva
CAS Number:
Specifications
- Particle size:75 µm
- pH range:3 - 13
- Description:Capto™ Phenyl (high sub)
- Size:25 mL
- Matrix:Highly cross-linked agarose
- Ligand:Phenyl
- Storage temperature:4 - 30 °C
- Binding capacity:27 mg BSA/ml
- Cat. no.:CA97067-924
- Type:Liquid
- Supplier No.:17545101
Specifications
About this item
Capto™ Phenyl (high sub) is used in the capture and intermediate stages of protein purification
- Improved productivity and process economy in downstream operations
- Very high flow rates and large sample volume processing
- Excellent chemical stability
The combination of high capacity, narrow specification range, high flow rate and low backpressure consistently reduces process cycle times and increases productivity.
Customers Who Bought This Also Bought