Phenyl Sepharose™ High Performance Hydrophobic Interaction Chromatography Media, Cytiva
Catalog #: CA97067-872
Supplier: Cytiva
CAS Number:
Specifications
- Particle size:34 µm
- pH range:3 - 13
- Description:Phenyl Sepharose™ High Performance
- Size:75 mL
- Matrix:Cross-linked agarose, spherical
- Ligand:Phenyl
- Storage temperature:4 - 30 °C
- Cat. no.:CA97067-872
- Type:Liquid
- Supplier No.:17108201
Specifications
About this item
Phenyl Sepharose High Performance is an aromatic hydrophobic interaction chromatography (HIC) medium, designed for intermediate and polishing step purification steps when high resolution has priority.
- Aromatic HIC medium designed for intermediate purification and polishing steps
- Selectivity similar to Phenyl Sepharose 6 Fast Flow (low sub) but higher resolution due to the smaller 34 µm bead size
- BioProcess medium supported for industrial applications and well-established in approved processes
- The hydrophilic nature of the base matrix ensures low levels of non-specific binding
- Stable in commonly used aqueous buffers
Customers Who Bought This Also Bought