About this item
Butyl Sepharose 4 Fast Flow is part of the Sepharose Fast Flow HIC platform, which has been an industrial standard for HIC chromatography during recent decades
- Standard aliphatic HIC media based on Sepharose Fast Flow base matrix derivatized via uncharged, chemically-stable ether linkages
- Optimized for the separation of larger proteins in capture and intermediate purification steps
- The hydrophilic nature of the base matrix ensures low levels of non-specific binding
BioProcess medium supported for industrial applications and well-established in approved processes. Butyl Sepharose 4 Fast Flow is available in a range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development. As member of the BioProcess media range, Butyl Sepharose 4 Fast Flow meets industrial demands with security of supply and comprehensive technical and regulatory support.
Butyl Sepharose 4 Fast Flow is based on cross-linked 4% agarose. The aliphatic ligand is immobilized to the base matrix with an ether linkage. The ligand contains no charged groups, making true hydrophobic interaction chromatography possible, without interfering ionic effects.
Related Information
Certificates
Search by Lot Number17098001_DATA FILE 1.pdf 17098001_DATA FILE 2.pdf 17098001_DATA FILE.pdf 17098010_DATA FILE 1.pdf 17098010_DATA FILE.pdf
Datasheet
17098001_HAND BOOK.pdf 17098001_INSTRUCTIONS 1.pdf 17098001_INSTRUCTIONS 2.pdf 17098001_INSTRUCTIONS 3.pdf 17098001_INSTRUCTIONS 4.pdf