About this item
Phenyl Sepharose 6 Fast Flow is a well established, standard aromatic hydrophobic interaction chromatography (HIC) resin for capture and intermediate purification requiring low to resin hydrophobicity
- Standard aromatic HIC resins based on Sepharose Fast Flow base matrix derivatized via uncharged, chemically-stable ether linkages.
- Designed for initial and intermediate purification requiring low to medium hydrophobicity
- Two levels of substitution help find optimal selectivity and binding capacity for a given application
- BioProcess resin supported for industrial applications and well-established in approved processes
- The hydrophilic nature of the base matrix ensures low levels of non-specific binding.
Phenyl Sepharose 6 Fast Flow is part of the Sepharose Fast Flow HIC platform, which has been an industrial standard for HIC chromatography during recent decades. Phenyl Sepharose 6 Fast Flow is composed of cross-linked, 6% agarose beads modified with standard aromatic phenyl groups via uncharged, chemically-stable ether linkages. Two levels of ligand substitution degree help to find the optimal selectivity and binding capacity for a given application.
Phenyl Sepharose 6 Fast Flow is available in a range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development. As member of the BioProcess resins range, Phenyl Sepharose 6 Fast Flow meets industrial demands with security of supply and comprehensive technical and regulatory support.
Related Information
Certificates
Search by Lot Number17097305_DATA FILE 1.pdf 17097305_DATA FILE 2.pdf 17097305_DATA FILE 3.pdf 17097305_DATA FILE.pdf 17097310_DATA FILE.pdf
Datasheet
17097305_HAND BOOK.pdf 17097305_INSTRUCTIONS 1.pdf 17097305_INSTRUCTIONS 2.pdf 17097305_INSTRUCTIONS 3.pdf 17097305_INSTRUCTIONS 4.pdf