HiPrep Phenyl HP Columns
Supplier: Cytiva
Certificates
About this item
HiPrep 16/10 Phenyl HP is a prepacked column designed for preparative hydrophobic interaction chromatography (HIC) with good resolution.
- Aromatic HIC resin for intermediate purification and polishing
- Selectivity similar to Phenyl Sepharose 6 Fast Flow (Low Sub) but higher resolution due to the smaller 34 µm bead size
- Provides fast, easy high-capacity separations in a convenient format
Phenyl Sepharose HP chromatography resin is based on a matrix of highly cross-linked agarose beads, which have been derivatized with phenyl groups. These are attached to the agarose matrix via uncharged, chemically stable ether linkages. The result is a truly hydrophobic resin with no ionic properties.
Phenyl Sepharose HP chromatography resin is based on a matrix of highly cross-linked agarose beads, which have been derivatized with phenyl groups. These are attached to the agarose matrix via uncharged, chemically stable ether linkages. The result is a truly hydrophobic resin with no ionic properties.
Specifications
- Catalog No:
- CA30621-420
- Max. pressure:
- 3 bar [0.3 MPa] (44 psi)
- pH range:
- 3 - 13
- Storage:
- 4...30 °C, 20% Ethanol, 0.2 M Sodium Acetate
Specifications
Related Information
Certificates
Search by Lot NumberCytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
Customers Who Bought This Also Bought