Glutathione Sepharose™ High Performance Affinity Chromatography Media, Cytiva
Supplier: Cytiva
Certificates
About this item
Glutathione Sepharose High Performance is used for the purification of GST-tagged proteins from bacterial lysates and other S-transferases or glutathione-dependent proteins.
- 34 µm bead size for improved efficiency and resolution
- Fast, one-step purification
- Mild conditions during the purification process preserves protein antigenicity and function
GST-tagged proteins are eluted under mild, nondenaturing conditions using reduced glutathione. The purification process preserves protein antigenicity and function. Fast, one-step purification of GST-tagged proteins and other glutathione S-trasferase or glutathione-dependent proteins.
Related Information
Certificates
Search by Lot Number17527901_DATA FILE 1.pdf 17527901_DATA FILE.pdf 17527902_DATA FILE 1.pdf 17527902_DATA FILE.pdf
Datasheet
17527901_HAND BOOK 1.pdf 17527901_HAND BOOK.pdf 17527901_INSTRUCTIONS.pdf 17527902_HAND BOOK 1.pdf 17527902_HAND BOOK.pdf
Manual
More
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
Customers Who Bought This Also Bought