Ni Sepharose™ High Performance Affinity Chromatography Media, Cytiva
Supplier: Cytiva
Certificates
About this item
Ni Ni Sepharose High Performance affinity media for high-resolution, high capacity purification of histidine-tagged proteins by immobilized metal ion affinity chromatography (IMAC).
- High-performance purification of histidine-tagged proteins
- Negligible leakage of Ni₂₊ ions
- High binding capacity, at least 40 mg/ml medium
- Compatible with a wide range of reducing agents, detergents, denaturants and other additives
- 34 µm bead size for improved resolution
Ni Sepharose High Performance consists of highly cross-linked agarose beads to which a chelating group has been coupled. This chelating group is precharged with Ni₂₊, which will selectively retain proteins with exposed histidine groups. The high stability and broad compatibility of Ni Sepharose High Performance maintains biological activity and increases product yield, at the same time as it greatly expands the range of suitable operating conditions. Chelating agents such as EDTA, EGTA , citrate should not be used.
Related Information
Certificates
Search by Lot Number17526801_DATA FILE 1.pdf 17526801_DATA FILE.pdf 17526802_DATA FILE 1.pdf 17526802_DATA FILE.pdf
Datasheet
17526801_HAND BOOK 1.pdf 17526801_HAND BOOK.pdf 17526801_INSTRUCTIONS.pdf 17526802_HAND BOOK 1.pdf 17526802_HAND BOOK.pdf
Manual
More
CA95055-838L_10292841
Conformity declaration
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
Customers Who Bought This Also Bought