Capto™ Blue Affinity Chromatography Media, Cytiva
Catalog #: CA97067-922
Supplier: Cytiva
CAS Number:
Specifications
- Particle size:75 µm
- pH range:2 - 13
- Description:Capto™ Blue
- Size:25 mL
- Matrix:Highly cross-linked agarose
- Ligand:Cibacron Blue
- Storage temperature:2 - 8 °C
- Binding capacity:>24 mg HSA/ml
- Cat. no.:CA97067-922
- Type:Liquid
- Supplier No.:17544801
Specifications
About this item
Capto Blue is an affinity medium for the capture of albumin, purification of enzymes, and recombinant proteins at laboratory and process scales.
- Excellent chemical stability ensures tolerance of harsh solvents used in repeated cleaning-in-place and sanitization procedures
- Capto Blue can be autoclaved repeatedly
- Highly rigid agarose base matrix allows high flow rates and processing of large sample volumes with no reduction in binding capacity
- Ligand functionality may be modified through the use of appropriate buffer salts and buffer conductivity to increase selectivity for desired targets
Excellent choice for the removal or purification of proteins at both laboratory and process scales.
Customers Who Bought This Also Bought