His Mag Sepharose Ni Magnetic Beads, Cytiva
Supplier: Cytiva
Certificates
About this item
His Mag Sepharose® Ni are magnetic beads designed for rapid, small-scale purification and screening of histidine-tagged proteins from different sources.
- Easy-to-use, visible magnetic beads ensures reliable collection of bound histidine-tagged proteins in purification procedures
- High purity and yield
- Easy parallel screening with high repeatability
- High density of beads ensures rapid capture from magnetic devices
- Scalable-simple capture of target proteins from small µl to high ml sample volumes
Lower concentrations of histidine-tagged proteins are commonly used to wash resins prior to elution. Mag Sepharose® format has excellent properties for small-scale experiments.
Specifications
- Catalog No:
- CA89166-170
- CA89166-172
- Ligand:
- IMAC immobilized with Ni
- IMAC immobilized with Ni
- Matrix:
- Paramagnetic, spherical, highly cross-linked agarose particles
- Paramagnetic, spherical, highly cross-linked agarose particles
- Operating temperature:
- Room temperature
- Room temperature
- Particle size:
- 37 - 100 μm
- 37 - 100 μm
- Solubility:
- Liquid and suspension; easily soluble in cold water and hot water
- Liquid and suspension; easily soluble in cold water and hot water
- Storage conditions:
- 4...30 °C, 20% ethanol, 5% medium slurry
- 4...30 °C, 20% ethanol, 5% medium slurry
Specifications
Related Information
Customers Who Bought This Also Bought