PD MidiTrap G-10 Columns, Cytiva
Catalog #: CA89128-872
Supplier: Cytiva
Specifications
- Capacity:5.3 ml
- Description:PD MidiTrap G-10
- Wetted materials:No
- Storage:4 to 30°C, 20% Ethanol
- Column Hardware:PP
- Bed Dimensions:13×40 mm
- Cat. no.:CA89128-872
- Sample Volume:400 - 1000 μl
- Supplier No.:28-9180-11
Specifications
About this item
PD MidiTrap G-10 columns are prepacked, single-use gravity columns for buffer exchange and cleanup of biological samples, for example peptides, small proteins, and oligosaccharides.
- Easy cleanup of peptides, small proteins, or carbohydrates larger than Mr 700, before downstream analysis such as liquid chromatography, electrophoresis, and MS
- Efficient removal of contaminants such as salts, dyes, and radioactive labels
- High desalting capacity
- Preparation of samples in the volume range 400 to 1000 μl
PD MidiTrap G-10 gravity columns give fast and simple desalting and buffer exchange without any need for a purification system, PD MiniTrap G-10 columns are prepacked and designed for rapid, convenient sample clean-up of small proteins, peptides, oligo-saccharides and other small biomolecules (>700 Mr).
For smaller scale please use PD MiniTrap G-10 columns (300 μl).
Delivery: Instructions are supplied with the columns.
Customers Who Bought This Also Bought