Buffer Reservoir, Cytiva
Catalog #: CA95017-305L
Supplier: Cytiva
Specifications
- Description:LabMate PD-10 Buffer Reservoir
- Supplier No.:18-3216-03
Specifications
About this item
Fast, affordable, high-recovery desalting and buffer exchange, as well as removal of low-molecular weight compounds packed with Sephadex G-25 Medium, LabMate buffer reservoir makes it easier and more convenient to equilibrate PD-10 columns.
- Easy to use and equilibrate, apply sample, elute
- Buffer reservoir for easy equilibration
- No wetted part
Packaging: Pack size 10
Customers Who Bought This Also Bought