To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:T4 DNA Ligase
- Cat. no.:CA95040-312L
- No. of Reactions:50
- Supplier No.:27-0361-01
Specifications
About this item
T4 DNA ligase catalyses the formation of a phosphodiester bond between the 5'-phosphoryl group and the 3'-hydroxyl group of two double-stranded DNA fragments. ATP is required for this reaction.
- For covalently linking DNA fragments in as little as 30 minutes
- Single dose, ambient-temperature-stable reaction mixes
Catalyzes the formation of a phosphodiester bond between the 5'-phosphoryl group and the 3'-hydroxyl group of two double-stranded DNA fragments. ATP is required for this reaction. Ligation of blunt-ended DNA is greatly stimulated by polyethylene glycol. Applications include self-circularization of linear DNA prior to transformation, ligation of insert to vector, and ligation of synthetic linkers to blunt-ended DNA. Each predispensed, single-reaction, ambient-temperature-stable tube contains a glassified reaction mix which, when reconstituted to a final reaction volume of 20 µl, yields a solution of reaction buffer, T4 DNA ligase, and ATP.
Storage conditions: Room Temperature