Specifications
- Temperature control range:ambient + 5°C to 60 °C
- Description:Quad Microplate Shaker-Incubator
- Power:5A
- Electrical:12V DC
- Temperature uniformity:± 0.2°C (at 37°C)
- Orbit:2 mm
- Shaking speed:250 to 1200 RPM
- Temperature setting:+25 to 60°C
- Timer:1 min. to 96 hrs. (with auto-off and audible alarm)
- Cat. no.:97025-554
- Supplier No.:PHMP-4 USA
Specifications
About this item
Excellent temperature uniformity across the platform/microplates (due to the PID bi-directional heating system, from both below the plates and above the plates in the lid) combined with variable speed and variable temperature produces the ideal thermoshaker for microplate incubations.
The heated lid completely covers the flat heating plate to provide bi-directional heating and excellent temperature stability and uniformity, whilst preventing condensation build up. The combination of bi-directional heating and powerful orbital shaking capability produces the maximum yield from incubations. It is very easy to operate, with simple set-up of temperature, shaking speed and time via push buttons and the clear 2-line LCD status display.
The PHMP microplate shaker–incubator has the following features: temperature setting range: + 25°C to 60°C; temperature stability ± 0.1°C; temperature uniformity ± 0.5°C due to the bi-directional heating system (platform and lid); shaking speed: 250 to 1200rpm; rapid heat-up; continuous or timed operation, with alarm buzzer and automatic switch-off; choice of two models with capacity for two (PHMP) or four (PHMP-4) 96- or 384-well microplates.
A versatile product for use in immunochemical reactions, enzyme and protein analysis and molecular biology techniques such as micro array analysis.