PHMP Twin Microplate Shaker-Incubator Thermoshaker, Grant Instruments
Catalog #: 97025-552
Supplier: GRANT USA INC.
Specifications
- Temperature control range:ambient + 5°C to 60 °C
- Description:Twin Microplate Shaker-Incubator
- Power:5A
- Electrical:12V DC
- Temperature uniformity:± 0.2°C (at 37°C)
- Orbit:2 mm
- Shaking speed:250 to 1200 RPM
- Temperature setting:+25 to 60°C
- Timer:1 min. to 96 hrs. (with auto-off and audible alarm)
- Cat. no.:97025-552
- Supplier No.:PHMP USA
Specifications
About this item
Excellent temperature uniformity across the platform/microplate (patented bi-directional heating of microplates) combined with variable speed and variable temperature produces the ideal thermoshaker for microplate incubations
- Temperature setting range: +25°C to 60°C
- Stability ±0.1°C, uniformity ±0.2°C due to the bi-directional heating system (platform and lid)
- Shaking speed: 250 to 1200 rpm
- Rapid heat-up
- Continuous or timed operation, with alarm buzzer and automatic switch-off facility
- Patented dual heating of microplates
A versatile product for use in immunochemical reactions, enzyme and protein analysis and molecular biology techniques such as micro array analysis.
Can be used with all types of standard depth microplates and offers three devices in one for maximum versatility and efficiency; A microplate thermoshaker, compact benchtop incubator without shaking, and a microplate shaker in cold or ambient temperatures.
Certifications: CE approved.
Ordering information: Supplied complete with instructions, and 12V power supply. 2 Year warranty.
Customers Who Bought This Also Bought