High Five General Purpose Latex Gloves, Powder-Free, Microflex®, Ansell
Supplier: Microflex
Certificates
About this item
An economical glove for non-medical applications.
- Textured for enhanced gripping power
- Powder-free
General-purpose powder-free latex glove for applications where medical-grade protection is not required.
Specifications
- Catalog No:
- CA89427-464
- CA89427-468
- Disposable / reusable:
- Disposable
- Disposable
- Latex/Latex-free:
- Latex
- Latex
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Cuff thickness:
- 3.5 mil
- 3.5 mil
- Finger thickness:
- 5.5 mil
- 5.5 mil
- Palm thickness:
- 4.5 mil
- 4.5 mil