Order Entry
ContactUsLinkComponent
PIG® Oil-Only Spill Kit in 30-Gallon Overpack Salvage Drum, New Pig
PIG® Oil-Only Spill Kit in 30-Gallon Overpack Salvage Drum, New Pig
Catalog #: 56620-374
Supplier:  New Pig
PIG* Spill Kit in a 30-gallon Overpak Salvage Drum
PIG® Oil-Only Spill Kit in 30-Gallon Overpack Salvage Drum, New Pig
Catalog #: 56620-374
Supplier:  New Pig

Specifications

  • Capacity:
    Up to 79.5 L (21 gal.)
  • Cat. no.:
    56620-374
  • Supplier No.:
    KIT436

Specifications

About this item

Overpack drum container is UN Rated for shipping waste after oil spill cleanup

  • Overpack is X-rated in Packing Groups I, II and III for shipping spill cleanup waste by land, sea or rail
  • Lightweight, 100% copolymer container resists chemicals and keeps contents clean and dry
  • Leakproof, twist-on lid is notched for easy removal or tightening with a 2×4 or pole
  • Large, color-coded label makes identification fast and accurate
  • Ledges molded into container make kit easy to move by forklift
  • Tamperproof seals help prevent pilfering of spill response supplies

Overpack is X-rated in Packing Groups I, II and III for shipping spill cleanup waste by land, sea or rail. Lightweight, 100% copolymer container resists chemicals and keeps contents clean and dry. Leakproof, twist-on lid is notched for easy removal or tightening with a 2x4 or pole. Large, color-coded label makes identification fast and accurate. Ledges molded into container make kit easy to move by forklift. Tamperproof seals help prevent pilfering of spill response supplies.

No wheels. UN1H2/X100/S.

Neatly prepacked container speeds access during a spill emergency. PIG Oil-Only Booms stop spreading spills; PIG Stat-Mat and Skimmers absorb quickly. Absorbents meet NFPA 99 standards for static decay to allow use with fuels and flammables

Customers Who Bought This Also Bought