Order Entry
ContactUsLinkComponent
PIG® Burpless® One-Hand-Sealable Steel Drum Funnel, New Pig
PIG® Burpless® One-Hand-Sealable Steel Drum Funnel, New Pig
Catalog #: 89073-811
Supplier:  New Pig
Burpless Drum Funnel - Yellow
PIG® Burpless® One-Hand-Sealable Steel Drum Funnel, New Pig
Catalog #: 89073-811
Supplier:  New Pig

Specifications

  • Description:
    PIG® Burpless® one-hand-sealable steel drum funnel
  • Dimensions:
    28.6 x 38.1 x 33 cm (11.25 x 15 x 13")
  • Cat. no.:
    89073-811
  • Supplier No.:
    DRM1125-YW-NPT

Specifications

About this item

This funnel with latching handle easily opens and closes lid in a simple, single-handed operation.

  • Lockable to prevent unauthorized access
  • Suitable for 5 to 55 gal. steel drums and pails with 2" NPT
  • Durable, chemical-resistant powder coat helps ensure long service life

Unique burpless design vents during pouring for fast, smooth flow with no dangerous splashback. Lets you pour without venting through 3/4" bung, a possible closed container reg violation if left open. Nitrile gasket and locking mechanism, completely sealed lid to help reduce vapor emissions.

Secure closure helps you meet hazardous waste and closed container regs, but opens and closes instantly.

BodyTextAccessories: The 6" mesh tube extinguishes sparks and flames. The brass composition is non corrosive and chemical resistant. FM approved for use in temporary drum fills.
Customers Who Bought This Also Bought