Order Entry
ContactUsLinkComponent
PYR Reagent, Hardy Diagnostics
PYR Reagent, Hardy Diagnostics
Catalog #: CA89407-840
CAS Number:  
PYR Reagent
PYR Reagent, Hardy Diagnostics
Catalog #: CA89407-840
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Packaging:
    Dropper Bottle
  • Size:
    15 mL
  • Storage temperature:
    2-8°C
  • Cat. no.:
    CA89407-840
  • Supplier No.:
    Z127

Specifications

About this item

For use in conjunction with PYR testing for the detection of pyroglutamate aminopeptidase found in group A streptococci Streptococcus pyogenes) and group D enterococci

The PYR Reagent combines with beta-naphthyl amide released during pyroglutamate aminopeptidase (PYR) activity to form a pink to red pigment. See Z75 for the complete kit.

Customers Who Bought This Also Bought