Crystal violet advanced for Gram staining
Catalog #: CA89405-046
Supplier: HARDY DIAGNOSTICS CA
CAS Number: 548-62-9
Specifications
- Size:3.79 L
- Cat. no.:CA89405-046
- Supplier No.:C128A
Specifications
Specification Test Results
- Crystal violet advanced for Gram staining:
Specification Test Results
About this item
Synonyms: Methylrosanilide chloride,Dimethyl((di(4-(dimethylamino)phenyl)methylen)-2,5-cyclohexadien-1-yliden)ammoniumchlorid,Viocid,Hexamethyl Violet,Basic Violet 3,Methyl violet 10B,Gentiaverm,CI 42555,4-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]dimethylammonium chloride,Hexamethyl-p-rosaniline chloride,Vianin,Gentian violet,N-(4-(Bis(4-(dimethylamino)phenyl)methylene)cyclohexa-2
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 189 to 194 °C
:Flash Pt:
:Flash Pt:
:M.W. 407.99 g/mol
:Crystal violet
Formula:C₂₅H₃₀ClN₃
MW:407.99 g/mol
MDL Number:MFCD00011750
CAS Number:548-62-9
Customers Who Bought This Also Bought