To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Mylar® zip-pouch
- Size:100 g
- Storage temperature:–2-30°C
- Cat. no.:CA89406-548
- Supplier No.:C7830
Specifications
About this item
CRITERION™ Cellobiose is primarily used in preparing microbiological culture media
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
For fermentation tests it is usually added at a final concentration of 0.5 to 1.0%. CRITERION™ Dehydratedculture media with engineered container.