About this item
Food grade high density polyethylene lab cart in 12 colors with built-in tray, slides under counters, benchtops and fume hoods.
- Available in 13 colors
- Fits comfortably under tables, counters, and benches to fill unused space (27 inches high)
- Non-metal rust proof polymer construction performs well in harsh, humid, corrosive environments
- Decontaminate with bleach and aqueous solutions
- 4 full-extension drawers
- 1 key locks all drawers
- 1 piece body construction
- Built-in tray top and handle for push pull control
- Chemically resistant
- 150 pound weight capacity
- Fully assembled
- American made by TrippNT
- Total replacement Lifetime Guarantee
The Compact Drawer Cart creates storage for; glassware, tools, utensils, reagents, chemicals, pens, pippettes, tweezers, log books, documents, bottles and other laboratory supplies. Features a 12-3/4 by 3-1/4 by 15-5/8 inch built-in Tray Top.
Features 4 full-extension drawers (pulls all the way out to fully access drawer contents) that are 11-1/2 inches wide by 4-1/2 inches high and 15-1/2 inches deep. Clearance from bottom of drawer to drawer above is 4-1/2 inches.
Industrial drawer slides keep drawer closed while cart is moving. Stong, ergonomic, high density polyethylene handles.
Perfect for labs, clinics, hospitals, doctor's offices and similar industries to add extra work space, storage, and mobility. The TrippNT Compact Drawer Cart is built for small spaces and fits under benches, counters, desks, tables, or fume hoods.
Specifications
- Catalog No:
- 89408-466
- Material:
- Food Grade High Density Polyethylene, Natural White; ABS (Acrylonitrile butadiene styrene) drawers; PVC drawer fronts
- Storage capacity:
- 2.2 cu. ft.
- Caster type:
- 3" Diameter Total Locking Polypropylene Casters (Locks rotation and swivel)
- Drawer dimensions:
- 111/2" x 41/2" x 151/2", Internal
- Tray dimensions:
- 123/4" x 31/4" x 155/8", Top