Suture Cart with Bulk Storage Area, Tall Slanted, TrippNT
Supplier: TrippNT
Certificates
About this item
White Tall Slanted Suture Cart with Bulk Storage Area is able to hold up to 48 standard suture boxes on 8 angled shelves for easy viewing and retrieval.
- Holds 48 standard suture boxes for easy retrieval
- Keeps suture boxes at a secure 45 degree angle
- Bulk storage area
- Bright white PVC and clear acrylic construction
- 4 door colors
- 3 inch casters (2 locking)
- Fully assembled, ready to use out of the box
- In stock and ready to ship
- American made by TrippNT
- Total Replacement Lifetime Guarantee
The large bulk storage area is perfect for storing additional boxes of sutures, bottles of irrigation water or sterile suturing kits. Sitting atop 4 smooth rolling casters, it can easily be moved from an operating room to an exam room to nearly any location where its services may be needed.
Constructed of durable PVC. Choose from four door colors.
Customers Who Bought This Also Bought