To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Size:500 Assays
- Storage conditions:–80 °C (Long Term)
- Application:Fluorescence Microscopy
- Cat. no.:89165-914
- Supplier No.:51025-K500
Specifications
About this item
The ER-ID® Green assay kit is specifically designed for use with RFP- or OFP-expressing cell lines, as well as cells expressing blue or cyan fluorescent proteins (BFPs, CFPs).
- Easily multiplexed with common fluorescent dyes and fluorescent proteins
- Highly resistant to photo-bleaching, concentration quenching and photo-conversion
- Stringently manufactured, to control and eliminate non-specific assay artifacts
ER-ID™ Green assay kit contains an endoplasmic reticulum-selective dye suitable for live cell, or detergent-permeabilized aldehyde-fixed cell staining. Micromolar concentrations of ER-ID™ Green dye are sufficient for staining mammalian cells, as validated with human cervical carcinoma cell line, human T-lymphocyte cell line, Jurkat, HeLa and human bone osteosarcoma epithelial cell line, U2OS.One important application of ER-ID™ Green dye is in fluorescence co-localization imaging with red fluorescent protein (RFP)- or orange fluorescent protein (OFP)-tagged proteins, a powerful approach for determining the targeting of molecules to intracellular compartments and for screening of their associations and interactions.
Additionally, the kit is suitable for use with live or post-fixed cells in conjunction with probes, such as labeled antibodies, or other fluorescent conjugates displaying similar spectral properties as Texas Red, or coumarin. A nuclear counterstain is provided to highlight this organelle as well.