To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Size:2x96 Tests
- Storage conditions:–80 °C (Long Term)
- Application:Flow Cytometry, Fluorescence Microscopy, Fluorescent Detection Microplate
- Cat. no.:89165-910
- Supplier No.:51023-KP002
- No. of Tests:70 Flow Cytometry Tests
Specifications
About this item
PROTEOSTAT® Protein aggregation assay provides a simple, homogenous assay format for monitoring peptide and protein aggregation in solution
- Validated for use with microplate or flow cytometry platform
- Extensively benchmarked with IgG
- Optimize buffers and excipients for protein formulation
- Performs with a wide pH and ionic strength range
- Quantitative detection of protein aggregates from visible to subvisible particles
The assay is capable of providing quantitative analysis of protein aggregation in a robust and high-throughput fashion (Z’ factor score >0.5). Lyophilized native and aggregated protein are provided as negative and positive controls for monitoring changes in protein aggregation status.
This is useful for defining optimal storage formulations for proteins, for screening of compounds that promote or inhibit protein aggregation and potentially for the sensitive measurement of molecular chaperone activity. The assay can be employed to streamline protein processing and formulation optimization procedures.