To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Fluorescein-12-UTP
- Size:250 nmoL
- Purity detail:Purified by ion-exchange chromatography.
- Purity:≥93% (HPLC)
- Concentration:10mM
- Formulation:Liquid. Solution in water.
- Cat. no.:89165-846
- Supplier No.:42834
Specifications
About this item
Fluorescein-12-UTP is ideal for in vitro transcription reactions catalyzed by T3, T7, or SP6 RNA polymerases and reverse transcriptase to produce fluorophore-labeled RNA transcripts. RNA-based probes can be used to identify complementary sequences by in situ hybridization to fixed cells and tissues by direct or indirect fluorescence detection or by membrane hybridization procedures.