To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Biotin-17-adenosine-5’-triphosphate, 250 nmol
- Size:250 nmoL
- Purity detail:Contains \<5% Bio-17-ADP.
- Purity:≥93% (HPLC)
- Concentration:10mM
- Formulation:Liquid. Solution in water.
- Supplier No.:42817
Specifications
About this item
Bio-17-ATP (Biotin-17-adenosine-5’-triphosphate) can replace ATP for in vitro transcription reaction catalyzed by T3, T7 or SP6 RNA polymerases. The biotin-labeled RNA transcripts produced by these reactions are suitable for a wide range of applications such as nucleic acid hybridization, sequencing, and genome analysis.
The transcription reaction produces multiple RNA copies of the DNA template(s) during a short incubation period. RNA probes offer higher target specificity and greater sensitivity than the corresponding DNA-DNA hybrids. The single-stranded RNA probes offer slectivity unavailable with double-stranded DNA probes, because they are strand-specific. Furthermore, RNA probes hybridize much more efficiently to target molecules than DNA probes because there is no self-hybridization. The biotin-labeled hybridized probes can be detected by a reporter molecule linked to streptavidin, avidin, or anit-biotin antibody. Such a complex can be detected directly, e.g. by excitation of a fluorophore conjugated to streptavidin, or indirectly, e.g. using an enzyme conjugate that can produce an insoluble colored precipitate.