To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Cyanine 3-UTP (Enhanced)
- Size:250 nmoL
- Purity detail:Purified by ion-exchange chromatography.
- Purity:≥93% (HPLC)
- Concentration:10mM
- Formulation:Liquid. Solution in water.
- Cat. no.:89165-754
- Supplier No.:42505
Specifications
About this item
Cyanine 3-UTP (Enhanced) can replace UTP as a substrate for T7 RNA polymerase in labeling systems that generate labeled probes through in vitro transcription. Probes generated by these methods are suitable for multicolor fluorescence analysis, specifically dual-color expression arrays in conjunction with cyanine 3-UTP .