Order Entry
ContactUsLinkComponent
BIOARRAY™ Single-round RNA Amplification and Biotin Labeling System (100 Reactions), Enzo Life Sciences
BIOARRAY™ Single-round RNA Amplification and Biotin Labeling System (100 Reactions), Enzo Life Sciences
Catalog #: 89165-746
Supplier:  Enzo Life Sciences
CAS Number:  
BIOARRAY™ Single-round RNA Amplification and Biotin Labeling System (100 Reactions), Enzo Life Sciences
Catalog #: 89165-746
Supplier:  Enzo Life Sciences
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    RNA Amplification and Biotin Labeling System (100 reactions)
  • Size:
    1 Pack
  • Storage conditions:
    −20 °C
  • Cat. no.:
    89165-746
  • Supplier No.:
    42421-100

Specifications

About this item

Improved data quality and analysis results through greater biotin incorporation.

The Labeling System provides an optimized protocol and reagents for the production of biotin-labeled antisense RNA (aRNA) from total cellular RNA samples (1µg-5µg) in less than 24 hours for array analysis

The 100-Reaction Single- Round RNA Amplification and Biotin Labeling System has been optimized for superior performance across several automated liquid handling platforms.The result is a comprehensive system that reduces variability while subsequently improving reproducibility, data quality and throughput for automated genomic environments.The complete system is composed of reagents for cDNA synthesis and in vitro transcription labeling and does not provide materials required for purification.

While cDNA quantities produced are sufficient for gene expression, manual sample preparation involving multiple enzymatic reactions and product purification steps are time consuming and error-prone.

Delivery: Kit contains 400 µL dNTP Mix (dN), 100 µL Promoter Primer (P), 200 µL First Strand Buffer (FB), 800 µL DTT (D), 100 µL Reverse Transcriptase (RT), 100 µL RNase Inhibitor (I), 500 µL DNA Polymerase (DP), 100 µL RNase H (RH), 1.5 mL Second Strand Buffer (SB), 600 µL IVT Reaction Buffer (RB), 600 µL 10X Biotin-Labeled Ribonucleotides (B), 600 µL Enhancer Cocktail (EC), 300 µL T7 RNA Polymerase (T7) and 13 mL Nuclease-free Water (W).
Customers Who Bought This Also Bought