To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Size:100 µg
- Tag sequence:HIS Tag
- Biological activity:Recombinant UbcH1 charges and supports ubiquitinylation in vitro. Typical enzyme concentration to support conjugation in vitro is 100nM to 1µM.
- Protein/peptide name:UbcH1
- Purity:>95% (SDS-PAGE)
- Concentration:1.0mg/ml
- Formulation:Liquid. In 20mM TRIS-HCl, pH 7.5, containing 0.5mM DTT.
- Cat. no.:89165-302
- Supplier No.:UW9020-0100
Specifications
About this item
Produced in E. coli.
Ubiquitinylation of proteins constitutes an important cellular mechanism for targeting short-lived proteins for degradation by the 26S proteasome. Three classes of enzymes are involved in the conjugation of ubiquitin to proteins. E1, the ubiquitin activating enzyme, activates ubiquitin through the ATP-dependent formation of a high-energy thiol ester bond between the carboxyl terminus of ubiquitin and the active-site cysteine within E1. This E1-activated ubiquitin is transferred to a cysteine residue of an E2, or ubiquitin-conjugating enzyme (UbC). E2 enzymes, either by themselves or in conjunction with E3 enzymes (ubiquitin ligases), then transfer ubiquitin to target proteins forming stable isopeptide bonds resulting in multi-ubiquitin chain formation. It is the diverse combinations of E2-E3 complexes which are thought to define substrate specificity. Among ubiquitin-conjugating enzymes, the mammalian ubiquitin conjugating enzyme UbcH2, also known as E2-25k, is unique in its ability to catalyse the synthesis in vitro of unanchored Lys48-linked poly-ubiquitin chains from mono- or poly-ubiquitin, E1, and ATP. The properties of truncated and chimeric forms of E2-25k suggest that the polyubiquitin chain synthesis activity of this E2 depends on specific interactions between its conserved 150-residue core domain and its unique 50-residue tail domain. Similarly E2-25k can catalyse the cyclisation of longer poly-ubiquitin chains, including tetra- and penta-ubiquitin. Although cyclic tri-ubiquitin resists hydrolysis by the PA700 or isopeptidase T deubiquitinylating enzymes, it can be disassembled to ubiquitin monomers by isopeptidase(s) in a red blood cell extract. Thus, if cyclic poly-ubiquitin forms in vivo, it will not accumulate as a dead-end product.