Specifications
- Description:Auto-Ubiquitinylation Kit
- Size:1 Kit
- Storage Temperature:-80 °C
- No. of Assays/Kit:10
- Supplier No.:UW0970-0001
Specifications
About this item
This E3 ligase auto-ubiquitinylation kit enables proteins to be tested for ubiquitin E3 ligase activity through assessment of their ability to under auto-ubiquitinylation
Utilizing the first three steps in the ubiquitin cascade the kit facilitates ubiquitinylation of known or putative E3 ligase enzymes followed by Western blot analysis using the highly sensitive reagents provided or using antibodies to the specific protein of interest (user supplied). A high integrity ubiquitin E3 ligase enzyme is also provided for use as a positive control.