Order Entry
ContactUsLinkComponent
Undeca-Ubiquitin (Linear)
Undeca-Ubiquitin (Linear)
Catalog #: 89165-228
Supplier:  Enzo Life Sciences
CAS Number:  
Undeca-Ubiquitin (Ub11) (Linear) 100 ug
Undeca-Ubiquitin (Linear)
Catalog #: 89165-228
Supplier:  Enzo Life Sciences
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Size:
    100 µg
  • Protein synonyms:
    UB11
  • Protein/peptide name:
    Undeca-Ubiquitin (linear)
  • Purity:
    >90% (SDS-PAGE)
  • Concentration:
    1mg/ml
  • Formulation:
    Liquid. In 50mM TRIS, pH 8.0, containing 50mM sodium chloride.
  • Cat. no.:
    89165-228
  • Supplier No.:
    UW0820-0100

Specifications

About this item

Produced in E. coli.

Linear polyubiquitin chains are useful tools for investigating, amongst other things, the specificity and reactivity of deubiquitinylating enzymes (DUBs) and the recognition and interaction of linear polyubiquitin modified proteins with by ubiquitin binding domain (UBDs) containing proteins.

Customers Who Bought This Also Bought