Order Entry
ContactUsLinkComponent
Human Recombinant Procathepsin K (from Insect Cells)
Human Recombinant Procathepsin K (from Insect Cells)
Catalog #: 89164-096
Supplier:  Enzo Life Sciences
CAS Number:  
Cathepsin K Proenzyme (Rec.) 10 ug
Human Recombinant Procathepsin K (from Insect Cells)
Catalog #: 89164-096
Supplier:  Enzo Life Sciences
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    Insect cells
  • Species:
    Human
  • Size:
    10 µg
  • Protein/peptide name:
    Procathepsin K
  • Purity:
    >95% (SDS-PAGE)
  • Cat. no.:
    89164-096
  • Supplier No.:
    SE367-0010

Specifications

About this item

Produced in insect cells. Full length glycosylated human procathepsin K.

Recombinant glycosylated procathepsin K (also known as cathepsin O or cathepsin O2) cloned from human cDNA, expressed in insect cells, and purified as full-length proenzyme. Cathepsin K, a member of the papain family of cysteine proteases, acts upon proteins such as collagen, AL amyloid, kinin, and elastin. It is a lysosomal protease expressed primarily in osteoclasts, but also in other cell types such as macrophages. It functions in bone remodeling and is implicated in disease states such as atherosclerosis, arthritis, and pycnodysostosis.

Customers Who Bought This Also Bought