Anti-Tyrosine Hydroxylase Rabbit Polyclonal Antibody
Catalog #: 89163-198
Supplier: Enzo Life Sciences
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Tyrosine Hydroxylase
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Mammals
- Antigen symbol:TH
- ImmunoChemistry:Yes
- Size:100 µL
- Western blot:Yes
- Form:Liquid. In 10mM HEPES, pH 7.5, containing 150mM NaCl, 10% BSA and 50% glycerol.
- Antigen synonyms:TH-4|tyrosine hydroxylase|EC 1.14.16.2|TYH|Tyrosine 3-hydroxylase|Tyrosine 3-monooxygenase|TY3H|TH isoform 3|TH isoform a
- Cat. no.:89163-198
- Supplier No.:SA497-0100
Specifications
About this item
Host: Rabbit
- For Immunocytochemistry, Immunohistochemistry, Western Blot
Recognizes mammalian tyrosine hydroxylase.
Type: Primary
Antigen: TYROSINE HYDROXYLASE
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mammals
Customers Who Bought This Also Bought