About this item
The proteasome is widely recognised as the central enzyme of non-lysosomal protein degradation. It is responsible for intracellular protein turnover and it is also critically involved in many regulatory processes and, in higher eukaryotes, in antigen processing. An enzymatic cascade is responsible for the attachment of multiple ubiquitin molecules to lysine residues of proteins targeted for degradation. The 26S proteasome is the key enzyme of the ubiquitin/ATP-dependent pathway of protein degradation.
The catalytic core of this unusually large (2000kDa, 450Å in length) complex (31 subunits) is formed by the 20S proteasome, a barrel shaped structure shown by electron microscopy to comprise of four rings each containing seven subunits. 20S Proteasomes degrade only unfolded proteins in an energy-independent manner, whereas 26S proteasomes degrade native and ubiquitinylated proteins in an ATP-dependent manner. The native protein substrates are recognised by subunits, some with ATP binding sites, of the outer 19S caps of the 26S proteasome. The 19S regulatory subunit recognizes ubiquitinated proteins and plays an essential role in unfolding and translocating targets into the lumen of the 20S subunit. A second activator which can associate with the 20S proteasome in the absence of ATP is known as PA28 or the 11S regulator. The pure PA28 activator is a complex of two alternating subunits, PA28α and PA28β, which share approximately 50% homology but also show considerable similarity (30-40%) to a nuclear protein of unknown function, the Ki autoantigen (recently referred to as PA28γ). These subunits, with an apparent relative molecular weight of approximately 29kDa, form ringlike heteromeric complexes of ~200kDa possibly with an α3β3 stoichiometry. Electron microscopic studies have shown PA28 to be a ring shaped particle which, like the 19S, caps the 20S proteasome, by binding to the α-rings, at both or either end. The complex may, however, be readily dissociated. The finding that PA28 modulates the proteasome-catalysed production of antigenic peptides presented to the immune system on MHC class I molecules indicates a cellular function of this activator in antigen processing. Several genetic diseases are associated with defects in the ubiquitin-proteasome pathway. Some examples of affected proteins include those linked to cystic fibrosis, Angelman's syndrome, and Liddle syndrome.
- For Western Blot
Recognizes human, mouse and rat proteasome activator 11S (alpha subunit).
Type: Primary
Antigen: PROTEASOME 11S ALPHA SUBUNIT
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 89162-044
- 89162-046
- Antigen symbol:
- Proteasome 11S Alpha Subunit
- Proteasome 11S Alpha Subunit
- Antigen name:
- Proteasome 11S Alpha Subunit
- Proteasome 11S Alpha Subunit
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Western blot:
- Yes
- Yes