To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:S. cerevisiae
- Antigen symbol:BLM10
- Size:25 µL
- Western blot:Yes
- Form:Liquid. Neat serum diluted in PBS containing 0.01% sodium azide.
- Cat. no.:89161-904
- Supplier No.:PW0570-0025
Specifications
About this item
Blm10 (mammalian homolog PA200) is a large 246kDa regulatory protein that binds to the 20S proteasome core particle (CP). Blm10 recognizes pre-activated open gate CP which is assumed to exist in equilibrium with inactive closed gate CP. Blm10 distinguishes gate conformations, regulates CP activation, and has been shown to be involved in CP maturation.
- For Western Blot
Recognizes Saccharomyces cervisiae Blm10.
Type: Primary
Antigen: BLM10
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Saccharomyces Cerevisiae